Learn More
Bachem Lys-(Des-Arg9)-Bradykinin Trifluoroacetate
SDP
Catalog No. 50259706
Encompass_Preferred
Lys-(Des-Arg9)-Bradykinin Trifluoroacetate, H-Lys-Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-OH, 71800-36-7, C50H73N13O11, Mr 1032.21, 100mg. (Des-Arg10)-Kallidin.
Catalog No. 50-259-706 Supplier Bachem Supplier No. 4026993.0100
Lys-(Des-Arg9)-Bradykinin Trifluoroacetate, H-Lys-Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-OH, 71800-36-7, C50H73N13O11, Mr 1032.21, 100mg. (Des-Arg10)-Kallidin.