Learn More
Medchemexpress LLC Lys-[des-Arg9]bradykinin trifluoroacetate | 2763588-90-3 | 98.9% | 1146.22 | C52H74F3N13O13 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY103295A10MG
Encompass_Preferred
Lys-[Des-Arg9]Bradykinin TFA is the trifluoroacetate salt of a bradykinin-derived peptide that acts as a potent and selective bradykinin B1 receptor agonist. It is supplied for use in receptor pharmacology, signaling studies, and in vitro assay development.
  • Potent, selective B1 receptor agonist (human Ki 0.12 nM, mouse Ki 1.7 nM, rabbit Ki 0.23 nM).
  • Trifluoroacetate (TFA) salt form for improved handling and solubility.
  • High purity suitable for in vitro pharmacology (98.9% by LC-MS/COA).
  • Molecular formula C52H74F3N13O13 and molecular weight 1146.22.
  • Available in small research quantities for assay and method development.
Catalog No. 50-002-44765