Learn More
Bachem Lys-(Des-Arg9,Leu8)-Bradykinin Trifluoroacetate
SDP
Catalog No. 50382592
Encompass_Preferred
Lys-(Des-Arg9,Leu8)-Bradykinin Trifluoroacetate, H-Lys-Arg-Pro-Pro-Gly-Phe-Ser-Pro-Leu-OH, 71800-37-8, C47H75N13O11, Mr 998.19, 25mg. (Des-Arg10,Leu9)-Kallidin. B1 bradykinin receptor antagonist.
Catalog No. 50-382-592 Supplier Bachem Supplier No. 4030394.0025
Add to Cart
Add to Cart
Lys-(Des-Arg9,Leu8)-Bradykinin Trifluoroacetate, H-Lys-Arg-Pro-Pro-Gly-Phe-Ser-Pro-Leu-OH, 71800-37-8, C47H75N13O11, Mr 998.19, 25mg. (Des-Arg10,Leu9)-Kallidin. B1 bradykinin receptor antagonist.