Learn More
Medchemexpress LLC Lys(MMT)-PAB-oxydiacetamide-PEG8-N3 | 1224601-12-0 | 95.2% | 1060.24 | C55H77N7O14 | 250 MG
SDP
Catalog No. 5000202411
Encompass_Preferred
Lys(MMT)-PAB-oxydiacetamide-PEG8-N3 is a cleavable antibody-drug conjugate linker reagent designed for bioconjugation and click chemistry. It features a Lys(MMT) protecting group, a p-aminobenzyl self-immolative spacer, an oxydiacetamide linkage, and a PEG8 spacer terminating in an azide to enable azide-alkyne click conjugation.
  • Cleavable PAB self-immolative spacer.
  • PEG8 spacer terminating in an azide for click conjugation.
  • Lys(MMT) protecting group for selective functionalization.
  • Purity approximately 95%.
  • Available in multiple milligram-scale pack sizes, including 250 mg.
Catalog No. 50-002-02411 Supplier Medchemexpress LLC Supplier No. HY131157250MG
Lys(MMT)-PAB-oxydiacetamide-PEG8-N3 is a cleavable antibody-drug conjugate linker reagent designed for bioconjugation and click chemistry. It features a Lys(MMT) protecting group, a p-aminobenzyl self-immolative spacer, an oxydiacetamide linkage, and a PEG8 spacer terminating in an azide to enable azide-alkyne click conjugation.
  • Cleavable PAB self-immolative spacer.
  • PEG8 spacer terminating in an azide for click conjugation.
  • Lys(MMT) protecting group for selective functionalization.
  • Purity approximately 95%.
  • Available in multiple milligram-scale pack sizes, including 250 mg.