Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lys-Nε-SPDB-DM4 | 1280215-91-9 | 99.8% | 1026.69 g/mol | C48H72ClN5O13S2 | 1 MG
SDP

Catalog No. 5000202683
Encompass_Preferred

Lys-Nε-SPDB-DM4 is a drug-linker conjugate that couples the tubulin inhibitor DM4 to a Lys-Nε-SPDB linker for use in antibody-drug conjugate (ADC) research. Supplied as a high-purity solid, it is intended for conjugation workflows, analytical characterization, and preclinical ADC development.

  • High purity (99.8%) solid for research use.
  • Contains DM4 tubulin inhibitor linked via Lys-Nε-SPDB for ADC conjugation.
  • Molecular weight approximately 1026.69 g/mol.
  • White to off-white powder, stable when stored sealed at 4°C.
  • Provided in small mg quantities suitable for synthesis and analytical workflows.

Catalog No. 50-002-02683 Supplier Medchemexpress LLC Supplier No. HY1415961MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lys-Nε-SPDB-DM4 is a drug-linker conjugate that couples the tubulin inhibitor DM4 to a Lys-Nε-SPDB linker for use in antibody-drug conjugate (ADC) research. Supplied as a high-purity solid, it is intended for conjugation workflows, analytical characterization, and preclinical ADC development.

  • High purity (99.8%) solid for research use.
  • Contains DM4 tubulin inhibitor linked via Lys-Nε-SPDB for ADC conjugation.
  • Molecular weight approximately 1026.69 g/mol.
  • White to off-white powder, stable when stored sealed at 4°C.
  • Provided in small mg quantities suitable for synthesis and analytical workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.