Learn More
Medchemexpress LLC Lys-SMCC-DM1 | 1281816-04-3 | 98.2% | 1103.71 g/mol | C53H75ClN6O15S | 1 MG
SDP
Catalog No. 5000245877
Encompass_Preferred
Lys-SMCC-DM1 is a lysine-conjugated SMCC-DM1 linker-payload metabolite used in antibody-drug conjugate (ADC) research. It serves as the active metabolite of the DM1 payload and is used as a linker-payload building block and analytical reference for ADC development, characterization, and stability studies. Molecular formula C53H75ClN6O15S; molecular weight 1103.71 g/mol. Store under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month).
  • Used as an ADC linker-payload building block and analytical reference.
  • Active DM1 metabolite that contributes to tubulin inhibition assays.
  • Purified small molecule suitable for conjugation workflows.
  • Stable under specified cold-storage conditions when stored under nitrogen.
  • Molecular characterization supports analytical method development.
Catalog No. 50-002-45877 Supplier Medchemexpress LLC Supplier No. HY1019821MG
Lys-SMCC-DM1 is a lysine-conjugated SMCC-DM1 linker-payload metabolite used in antibody-drug conjugate (ADC) research. It serves as the active metabolite of the DM1 payload and is used as a linker-payload building block and analytical reference for ADC development, characterization, and stability studies. Molecular formula C53H75ClN6O15S; molecular weight 1103.71 g/mol. Store under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month).
  • Used as an ADC linker-payload building block and analytical reference.
  • Active DM1 metabolite that contributes to tubulin inhibition assays.
  • Purified small molecule suitable for conjugation workflows.
  • Stable under specified cold-storage conditions when stored under nitrogen.
  • Molecular characterization supports analytical method development.