Learn More
TARGETMOL CHEMICALS INC Lys-SMCC-DM1 1MG
SDP
Catalog No. 502879824
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Lys-SMCC-DM1 (Lys-N-MCC-DM1) is an antibody-drug conjugate (ADC) targeting human epidermal growth factor receptor 2 (HER2). It contains the microtubule polymerization inhibitor DM1 as the active metabolite, which inhibits microtubule polymerization. It is commonly used in breast cancer research.
Catalog No. 50-287-9824 Supplier TARGETMOL CHEMICALS INC Supplier No. T119171MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Lys-SMCC-DM1 (Lys-N-MCC-DM1) is an antibody-drug conjugate (ADC) targeting human epidermal growth factor receptor 2 (HER2). It contains the microtubule polymerization inhibitor DM1 as the active metabolite, which inhibits microtubule polymerization. It is commonly used in breast cancer research.