Learn More
Selleck Chemical LLC Lys05 5mg 1391426-24-6
SDP
Supplier:  Selleck Chemical LLC S83695MG
Encompass_Preferred
Lys05 is a new lysosomal autophagy inhibitor which potently accumulates within and deacidifies the lysosome of both cells and tumors, resulting in sustained inhibition of autophagy and tumor growth. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8049