Learn More
CPC Scientific H-Pro-Glu-Pro-Ala-Lys-Ser-Ala-Pro-Ala-Pro-Lys-Lys(Ac)-Gly-Ser-Lys-Lys-Ala-Val-Thr-Lys-Ala-Gln-Gly-Gly-Lys(Biotin)-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST014B
Encompass
This peptide is the fragment of Histone H2B (1-22). It is acetylated at lysine 12 with a C-terminal GG linker followed by a biotinylated lysine. The histone H2B with acetylated at lysine 12 is a target for autoantibodies in systemic lupus erythematosus when it is apoptosis-induced.ONE-LETTER SEQUENCE: PEPAKSAPAPK-K(Ac)-GSKKAVTKAQ-GGK(Biotin)-NH2MOLECULAR FORMULA: MOLECULAR WEIGHT:2729.3STORAGE CONDITIONS: -20 5CRESEARCH AREA: MiscellaneousREFERENCES: Van Bavel, CC. et al. Mol Immunol 47, 2 (2009)
Catalog No. 50-210-9445