Learn More
CPC Scientific H-Ala-Arg-Thr-Lys-Gln-Thr-Ala-Arg-Lys-Ser-Thr-Gly-Gly-Lys(Ac)-Ala-Pro-Arg-Lys-Gln-Leu-Ala-Thr-Lys(Ac)-Ala-Gly-Gly-Lys(Biotin)-OH (trifluoroacetate salt) 0.25MG
SDP
Supplier:  CPC Scientific HIST035A
Encompass
This peptide is the fragment of Histone H3 (1-24) acetylated at Lys14 and Lys23, with a C-terminal GG linker followed by a biotinylated Lys. Lysine acetylation of histone H3 is associated with histone H4 methylation and transcriptional activation.ONE-LETTER SEQUENCE: ARTKQTARKSTGG-K(Ac)-APRKQLAT-K(Ac)-AGG-K(Biotin)MOLECULAR FORMULA: MOLECULAR WEIGHT:3107.7STORAGE CONDITIONS: -20 5CRESEARCH AREA: MiscellaneousREFERENCES: Garcia, B. et al. Nat Methods 4, 487 (2007)
Catalog No. 50-210-9469