Learn More
CPC Scientific H-Lys-Ser-Thr-Gly-Gly-Lys(Ac)-Ala-Pro-Arg-Lys-Gln-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST044A
Encompass
This peptide is the fragment of Histone H3 (9-19) acetylated at Lys14.ONE-LETTER SEQUENCE: KSTGG-K(Ac)-APRKQMOLECULAR FORMULA: C50H90N18O16MOLECULAR WEIGHT:1199.37STORAGE CONDITIONS: -20 5CRESEARCH AREA: MiscellaneousREFERENCES: Borra, MT. et al. Biochem. 43, 9877 (2004)
Catalog No. 50-210-9482