Learn More
CPC Scientific H-Lys-Ala-Ala-Arg-Lys(Ac)-Ser-Ala-Pro-Ala-Thr-Gly-Gly-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST060A
Encompass
This peptide is the fragment of Histone H3 (23-34) acetylate at Lys27. It is the least abundant isoform and is associated with histone deposition in replicating chromatin.ONE-LETTER SEQUENCE: KAAR-K(Ac)-SAPATGGMOLECULAR FORMULA: C48H85N17O16MOLECULAR WEIGHT:1156.3STORAGE CONDITIONS: -20 5CRESEARCH AREA: MiscellaneousREFERENCES: Garcia, B. et al. J Biol Chem 282, 7641 (2007); Schiltz, R. et al. J Biol Chem 274, 1189 (1999)
Catalog No. 50-210-9510