Learn More
CPC Scientific H-Ala-Pro-Arg-Lys-Gln-Leu-Ala-Thr-Lys-Ala-Ala-Arg-Lys(Me)-Ser-Ala-Pro-Ala-Thr-Gly-Gly-Val-Lys-Lys-Pro-His-Lys(Biotin)-NH2 (trifluoroacetate salt) 0.25MG
SDP
Supplier:  CPC Scientific HIST045A
Encompass
This peptide is the fragment of Histone H3 (15-39) monomethylated at Lys27 with a C-terminal followed by a biotinylated Lys.ONE-LETTER SEQUENCE: APRKQLATKAAR-K(Me)-SAPATGGVKKPH-K(Biotin)-NH2MOLECULAR FORMULA: MOLECULAR WEIGHT:2937.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: Miscellaneous
Catalog No. 50-210-9483