Learn More
CPC Scientific H-Lys-Ala-Ala-Arg-Lys(Me1)-Ser-Ala-Pro-Ala-Thr-Gly-Gly-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST061A
Encompass
This peptide is the fragment of Histone H3 (23-34) mono-methylated at Lys27.ONE-LETTER SEQUENCE: KAAR-K(Me1)-SAPATGGMOLECULAR FORMULA: MOLECULAR WEIGHT:1128.3STORAGE CONDITIONS: -20 5CRESEARCH AREA: Miscellaneous
Catalog No. 50-210-9511