Learn More
CPC Scientific H-Ala-Thr-Lys-Ala-Ala-Arg-Lys-Ser-Ala-Pro-Ala-Thr-Gly-Gly-Val-Lys(Me3)-Lys-Pro-His-Arg-Tyr-Arg-Pro-Gly-Gly-Lys(Biotin)-OH (trifluoroacetate salt) 0.25MG
SDP
Catalog No. 502109486
Encompass
This peptide is the fragment of Histone H3 (21-44) tri-methylated at Lys36, with an additional C-terminal Gly followed by a biotinylated Lys.ONE-LETTER SEQUENCE: ATKAARKSAPATGGV-K(Me3)-KPHRYRPG-GK(Biotin)MOLECULAR FORMULA: MOLECULAR WEIGHT:2959.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: Miscellaneous
Catalog No. 50-210-9486 Supplier CPC Scientific Supplier No. HIST047A
Add to Cart
Add to Cart
This peptide is the fragment of Histone H3 (21-44) tri-methylated at Lys36, with an additional C-terminal Gly followed by a biotinylated Lys.ONE-LETTER SEQUENCE: ATKAARKSAPATGGV-K(Me3)-KPHRYRPG-GK(Biotin)MOLECULAR FORMULA: MOLECULAR WEIGHT:2959.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: Miscellaneous