Learn More
CPC Scientific H-Ser-Thr-Gly-Gly-Val-Lys(Me3)-Lys-Pro-His-Arg-Tyr-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST065A
Encompass
This peptide is the fragment of Histone H3 (31-41) tri-methylated at Lys-36ONE-LETTER SEQUENCE: STGGV-K(Me3)-KPHRYMOLECULAR FORMULA: MOLECULAR WEIGHT:1271.4STORAGE CONDITIONS: -20 5CRESEARCH AREA: Miscellaneous
Catalog No. 50-210-9516