Learn More
CPC Scientific H-Gly-Ser-Arg-Ala-His-Ser-Ser-His-Leu-Lys(Me3)-Ser-Lys-Lys-Gly-Gln-Ser-Thr-Ser-Arg-His-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CANC014B
Encompass
p53K370Me3 is derived from amino acid residues 361-380 of human p53 tumor suppressor protein, with trimethylation at Lys370. Trimethylation at K370 does not occur in vivo but can be used as a control.ONE-LETTER SEQUENCE: GSRAHSSHL-K(Me3)-SKKGQSTSRHMOLECULAR FORMULA: MOLECULAR WEIGHT:2218.4STORAGE CONDITIONS: -20 5CRESEARCH AREA: CancerREFERENCES: Huang, J. et al. Nature 449, 105 (2007)
Catalog No. 50-210-8891