Learn More
CPC Scientific H-Gly-Ser-Arg-Ala-His-Ser-Ser-His-Leu-Lys(Me2)-Ser-Lys-Lys-Gly-Gln-Ser-Thr-Ser-Arg-His-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CANC011B
Encompass
p53K370Me2 is derived from amino acid residues 361-380 of human p53 tumor suppressor protein, with dimethylation at Lys370. This peptide promotes p53 interactions with both p21gene promoters and p53 binding protein 1 (53BP1), activating transcription of p53 target genesONE-LETTER SEQUENCE: GSRAHSSHL-K(Me2)-SKKGQSTSRHMOLECULAR FORMULA: MOLECULAR WEIGHT:2204.4STORAGE CONDITIONS: -20 5CRESEARCH AREA: CancerREFERENCES: Egorova, K. Biochem 75, 535 (2010)
Catalog No. 50-210-8885