Learn More
CPC Scientific Biotin-Lys-Gly-Gly-His-Leu-Lys-Ser-Lys-Lys-Gly-Gln-Ser-Thr-Ser-Arg-His-Lys-Lys(Ac)-Leu-Met-Phe-Lys-Thr-Glu-Gly-Pro-Asp-Ser-Asp-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CANC005B
Encompass
This peptide is tumor suppressor protein p53 (368-393). There is a biotinylated lysine follwed by a GG linker at the N-terminal and is acetylated at lysine 382. p53 is an ideal tumor suppressor gene because of its ability to control cell proliferation, and senescence. In response to cell stress, cell damage, or ectopic oncogene, p53 also mediates apoptosis.ONE-LETTER SEQUENCE: Biotin-KGGHLKSKKGQSTSRHK-K(Ac)-LMFKTEGPDSD-NH2MOLECULAR FORMULA: MOLECULAR WEIGHT:3481STORAGE CONDITIONS: -20 5CRESEARCH AREA: CancerREFERENCES: Ghosh, A. et al. Mol. Cell Biol. 24, 7987 (2004)
Catalog No. 50-210-8875