Learn More
CPC Scientific H-Ala-Arg-Thr-Lys(Me3)-Gln-Thr-Ala-Arg-Lys-Ser-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST022A
Encompass
This peptide is the fragment of Histone H3 (1-10) tri-methylated at Lys4.ONE-LETTER SEQUENCE: ART-K(Me3)-QTARKSMOLECULAR FORMULA: MOLECULAR WEIGHT:1188.3STORAGE CONDITIONS: -20 5CRESEARCH AREA: Miscellaneous
Catalog No. 50-210-9455