Learn More
CPC Scientific H-Ala-Arg-Thr-Lys(Me1)-Gln-Thr-Ala-Arg-Lys-Ser-Thr-Gly-Gly-Lys-Ala-Pro-Arg-Lys-Gln-Leu-Ala-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST028A
Encompass
This peptide is the fragment of Histone H3 (1-21) mono-methylated at Lys4.ONE-LETTER SEQUENCE: ART-K(Me1)-QTARKSTGGKAPRKQLAMOLECULAR FORMULA: MOLECULAR WEIGHT:2268.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: MiscellaneousREFERENCES: Ortega-Velazquez, R. et al. FASEB. 17, 1529 (2003)
Catalog No. 50-210-9461