Learn More
CPC Scientific H-Ala-Arg-Thr-Lys(Me2)-Gln-Thr-Ala-Arg-Lys-Ser-Thr-Gly-Gly-Lys-Ala-Pro-Arg-Lys-Gln-Leu-Ala-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502109462
Encompass
This peptide is the fragment of Histone H3 (1-21) di-methylated at Lys4.ONE-LETTER SEQUENCE: ART-K(Me2)-QTARKSTGGKAPRKQLAMOLECULAR FORMULA: MOLECULAR WEIGHT:2282.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Miscellaneous
Catalog No. 50-210-9462 Supplier CPC Scientific Supplier No. HIST029A
Add to Cart
Add to Cart
This peptide is the fragment of Histone H3 (1-21) di-methylated at Lys4.ONE-LETTER SEQUENCE: ART-K(Me2)-QTARKSTGGKAPRKQLAMOLECULAR FORMULA: MOLECULAR WEIGHT:2282.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Miscellaneous