Learn More
CPC Scientific H-Ala-Arg-Thr-Lys(Me3)-Gln-Thr-Ala-Arg-Lys-Ser-Thr-Gly-Gly-Lys-Ala-Pro-Arg-Lys-Gln-Leu-Ala-Thr-Lys-Ala-Ala-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST037B
Encompass
This peptide is the fragment of human histone H3 (1-25) trimethylated at Lys4. It shows cell state and lineage potential by differentiating genes that are expressed, poised for expression, or repressed, and also labels imprinting control regionsONE-LETTER SEQUENCE: ART-K(Me3)-QTARKSTGGKAPRKQLATKAA-NH2MOLECULAR FORMULA: MOLECULAR WEIGHT:2667.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: MiscellaneousREFERENCES: Midorikawa, K. et al. Biochem Biophys Res Commun 333, 1073 (2005)
Catalog No. 50-210-9473