Learn More
Abcam LYS6K2, S6 kinase inhibitor, 5MG
Catalog No. 502704849
Encompass_Preferred
MW 445.42 g/mol. Selective, cell-permeable S6 kinase inhibitor. Blocks phosphorylation of p70 ribosomal S6 kinase.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4849 Supplier Abcam Supplier No. AB1461995MG
Add to Cart
Add to Cart
MW 445.42 g/mol. Selective, cell-permeable S6 kinase inhibitor. Blocks phosphorylation of p70 ribosomal S6 kinase.
The product is subject to the following: Abcam Restricted Use Statement