Learn More
Bachem (Lys8)-Vasopressin
SDP
Supplier:  Bachem 4014772.0005
Encompass_Preferred
(Lys8)-Vasopressin, H-Cys-Tyr-Phe-Gln-Asn-Cys-Pro-Lys-Gly-NH2, 50-57-7, C46H65N13O12S2, Mr 1056.23, 5mg. Lypressin, Terlipressin (4-12), LVP. A compound with reduced vasopressor and antidiuretic activity, as compared to AVP.
Catalog No. 50-259-618