Learn More
CPC Scientific H-Ala-Arg-Thr-Lys-Gln-Thr-Ala-Arg- Lys(Ac)-Ser-Thr-Gly-Gly-Lys-Ala-Pro-Arg-Lys-Gln-Leu-Ala-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST031A
Encompass
This peptide is the residues 1-21 of the histone H3 acetylated at Lys9. It is highly localized to the 5? region of transcription start sites in human genes. This peptide is essential for transcription initiation and elongation.ONE-LETTER SEQUENCE: ARTKQTAR-K(Ac)-STGGKAPRKQLAMOLECULAR FORMULA: C96H174N36O29MOLECULAR WEIGHT:2296.7STORAGE CONDITIONS: -20 5CRESEARCH AREA: MiscellaneousREFERENCES: Midorikawa, K. et al. Biochem Biophys Res Commun 333, 1073 (2005)
Catalog No. 50-210-9465