Learn More
CPC Scientific H-Ala-Arg-Thr-Lys-Gln-Thr-Ala-Arg-Lys(Ac)-Ser-Thr-Gly-Gly-Lys-Ala- Pro-Arg-Lys-Gln-Leu-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST027A
Encompass
This peptide is the fragment of Histone H3 (1-20) acetylated at Lys9.ONE-LETTER SEQUENCE: ARTKQTAR-K(Ac)-STGGKAPRKQLMOLECULAR FORMULA: C93H169N35O28MOLECULAR WEIGHT:2225.56STORAGE CONDITIONS: -20 5CRESEARCH AREA: MiscellaneousREFERENCES: Borra, MT. et al. Biochem. 43, 9877 (2004)
Catalog No. 50-210-9460