Learn More
CPC Scientific H-Ala-Arg-Thr-Lys-Gln-Thr-Ala-Arg-Lys(Me3)-Ser-Thr-Gly-Gly-Lys-Ala-Pro-Arg-Lys-Gln-Leu-Ala-Thr-Lys-Ala-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502109468
Encompass
This peptide is the fragment of Histone H3 (1-24) tri-methylated at Lys9.ONE-LETTER SEQUENCE: ARTKQTAR-K(Me3)-STGGKAPRKQLATKAMOLECULAR FORMULA: MOLECULAR WEIGHT:2597STORAGE CONDITIONS: -20 5CRESEARCH AREA: Miscellaneous
Catalog No. 50-210-9468 Supplier CPC Scientific Supplier No. HIST034A
This peptide is the fragment of Histone H3 (1-24) tri-methylated at Lys9.ONE-LETTER SEQUENCE: ARTKQTAR-K(Me3)-STGGKAPRKQLATKAMOLECULAR FORMULA: MOLECULAR WEIGHT:2597STORAGE CONDITIONS: -20 5CRESEARCH AREA: Miscellaneous