Learn More
CPC Scientific H-Thr-Lys-Gln-Thr-Ala-Arg-Lys(Me1)-Ser-Thr-Gly-Gly-Lys-Ala-Pro-Arg-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST040A
Encompass
This peptide is the fragment of Histone H3 (3-17) mono-methylated at Lys-9.ONE-LETTER SEQUENCE: TKQTAR-K(Me1)-STGGKAPRMOLECULAR FORMULA: MOLECULAR WEIGHT:1600.8STORAGE CONDITIONS: -20 5CRESEARCH AREA: Miscellaneous
Catalog No. 50-210-9478