Learn More
CPC Scientific H-Ala-Arg-Thr-Lys-Gln-Thr-Ala-Arg-Lys(Me3)-Ser-Thr-Gly-Gly-Lys-Ala-Pro-Arg-Lys-Gln-Leu-Ala-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST032A
Encompass
This peptide is the fragment of Histone H3 (1-21) tri-methylated at Lys9.ONE-LETTER SEQUENCE: ARTKQTAR-K(Me3)-STGGKAPRKQLAMOLECULAR FORMULA: MOLECULAR WEIGHT:2296.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Miscellaneous
Catalog No. 50-210-9466