Learn More
BPS Bioscience Inc LysA Universal PARylation Assay Kit, Sandwich ELISA, 96-well plate, 100 pM to 20 nM PARylation range
SDP
Supplier:  BPS Bioscience Inc 82123
Encompass
The LysA Universal PARylation Assay Kit from BPS Bioscience is meticulously designed for the quantitative measurement of total poly ADP-ribosylation (PARylation) in cell extracts. This kit offers a convenient method for researchers to assess PAR levels, making it ideal for screening and determining the IC50 values of PARP or PARG inhibitors and validating their effects in cellular models. It specifically detects poly-ADP-ribosylation and is not designed for mono-(ADP-ribose) modifications.
  • Enables precise quantification of total PARylation levels using a PAR standard to establish a standard curve.
  • Supplied in a ready-to-use 96-well plate format with necessary reagents for streamlined workflows.
  • Demonstrates sensitivity in the 100 pM to 20 nM PARylation range.
  • Compatible for assessing the efficacy of PARP and PARG inhibitors.
  • Optimized for performance for up to 6 months when stored properly.
Catalog No. NC2680313