Learn More
Carbosynth LLC. Lysenin Antiserum (50 µl vial), 1ea, Lysenin is a sphingomyelin-binding protein and pore-forming toxin first isolated from the earthworm, Eisenia foetida.
SDP
Supplier:  Carbosynth LLC. NLY14802V50UL
Encompass_Preferred
Application: Our undiluted antisera are suitable for immuno-histochemical use, for application to radioimmunoassay (RIA), or other non-isotopicimmunoassay systems. These antisera are partially characterized by either of the following two methods: 1) (EIA) or 2) ELISA, Storage Temperature: 2-10°C
Catalog No. 50-168-6056