Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysine Acetate, Spectrum™ Chemical
SDP

Catalog No. 18620998 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-998 100 g
18-620-997 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-620-998 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. LY103100GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synonym: L-Lysine Acetate, L-Lysine Monoacetate, L-Lysine Acetate Salt

Lysine Acetate, also known as 2,6-Diaminohexanoic acid, is an essential amino acid which participates in hydrogen bonding and can be used as a base in catalysis. Its codons are AAA and AAG. Ungraded products supplied by Spectrum™ are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Chemical Identifiers

CAS 57282-49-2
Molecular Weight (g/mol) 206.24

Specifications

Quantity 100 g
Packaging Poly
Molecular Formula C8H18N2O4
MDL Number MFCD00039069
InChI Key RRNJROHIFSLGRA-JEDNCBNOSA-N
SMILES CC(O)=O.NCCCC[C@H](N)C(O)=O
IUPAC Name (2S)-2,6-diaminohexanoic acid; acetic acid
Grade Ungraded
Chemical Name or Material Lysine Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.