Learn More
Alkali Scientific Lysine Agar, 10 mL, 60 × 15 mm Plate, 10/Pk
SDP
Catalog No. 5000662090
Encompass_Preferred
Lysine Agar, 10 mL, 60 × 15 mm Plate, 10/Pk. This pre-prepared agar plate is ideal for detecting lysine decarboxylation, a key biochemical test for identifying specific bacterial species. It is designed for quick, accurate testing in microbiology labs, providing clear results for bacterial differentiation. The plate format ensures easy handling and convenient use, making it a valuable tool for laboratory analysis.
Catalog No. 50-006-62090 Supplier Alkali Scientific Supplier No. LYSAP00
Lysine Agar, 10 mL, 60 × 15 mm Plate, 10/Pk. This pre-prepared agar plate is ideal for detecting lysine decarboxylation, a key biochemical test for identifying specific bacterial species. It is designed for quick, accurate testing in microbiology labs, providing clear results for bacterial differentiation. The plate format ensures easy handling and convenient use, making it a valuable tool for laboratory analysis.