Learn More
Bioworld Lysine Iron Agar (LIA), 500 g
Catalog No. 50254327
Encompass_Preferred
Lysine Iron Agar (LIA)Used to detect the production of hydrogen sulfide and lysine decarboxylase by Enterobacteriaceae pH: 6.7Working Concentration: 34.5 g/LDehydrated Appearance: beige to greenish beige, homogeneous powderPrepared Appearance: purple agarSoluble in boiling waterContains:Pancreatic Digest of Gelatin Yeast Extract Dextrose L-Lysine Ferric Ammonium Citrate Sodium Thiosulfate Bromcresol Purple Agar
Catalog No. 50-254-327 Supplier Bioworld Supplier No. 306200451
Lysine Iron Agar (LIA)Used to detect the production of hydrogen sulfide and lysine decarboxylase by Enterobacteriaceae pH: 6.7Working Concentration: 34.5 g/LDehydrated Appearance: beige to greenish beige, homogeneous powderPrepared Appearance: purple agarSoluble in boiling waterContains:Pancreatic Digest of Gelatin Yeast Extract Dextrose L-Lysine Ferric Ammonium Citrate Sodium Thiosulfate Bromcresol Purple Agar