Learn More
Alkali Scientific Lysine Iron Agar Slant, 4mL, 13x100mm, Screw Cap Glass, 10/Pk
SDP
Catalog No. 5000662210
Encompass_Preferred
Lysine Iron Agar (LIA) Slants, 4mL, 13 × 100mm Glass Tubes, 10/Pk are differential culture media designed for the detection of lysine decarboxylation, lysine deamination, and hydrogen sulfide (H₂S) production in enteric bacteria. The medium is commonly used for identification and differentiation of organisms such as Salmonella and Shigella in clinical, food safety, and environmental microbiology laboratories. The 4mL slant format in screw cap glass tubes provides a compact and secure environment for microbial incubation and storage. Characteristic color reactions and precipitate formation help microbiologists interpret metabolic activity and identify bacterial species. Supplied as sterile ready to use slants, LIA supports reliable biochemical testing and organism differentiation in routine microbiological workflows.
Catalog No. 50-006-62210 Supplier Alkali Scientific Supplier No. LIAT00
Lysine Iron Agar (LIA) Slants, 4mL, 13 × 100mm Glass Tubes, 10/Pk are differential culture media designed for the detection of lysine decarboxylation, lysine deamination, and hydrogen sulfide (H₂S) production in enteric bacteria. The medium is commonly used for identification and differentiation of organisms such as Salmonella and Shigella in clinical, food safety, and environmental microbiology laboratories. The 4mL slant format in screw cap glass tubes provides a compact and secure environment for microbial incubation and storage. Characteristic color reactions and precipitate formation help microbiologists interpret metabolic activity and identify bacterial species. Supplied as sterile ready to use slants, LIA supports reliable biochemical testing and organism differentiation in routine microbiological workflows.