Learn More
Alkali Scientific Lysine Iron Agar Slant, 9mL, 13x100mm, Slip Cap, 10/Pk
SDP
Catalog No. 5000662143
Encompass_Preferred
Lysine Iron Agar (LIA) Slants, 9mL, 13 × 100mm Glass Tubes, 10/Pk are differential culture media used for detecting lysine decarboxylation, lysine deamination, and hydrogen sulfide (H₂S) production in enteric bacteria. The formulation supports identification and differentiation of organisms such as Salmonella and Shigella during microbiological analysis. The 9mL slant format provides extended media volume for laboratories that require longer incubation periods or larger inoculum s. Characteristic color changes and precipitate formation allow microbiologists to observe metabolic reactions and identify bacterial species. Packaged in sterile glass tubes, LIA slants provide a reliable and convenient solution for biochemical testing, bacterial differentiation, and routine microbiology workflows in clinical, food safety, and environmental laboratories.
Catalog No. 50-006-62143 Supplier Alkali Scientific Supplier No. LIAT01
Lysine Iron Agar (LIA) Slants, 9mL, 13 × 100mm Glass Tubes, 10/Pk are differential culture media used for detecting lysine decarboxylation, lysine deamination, and hydrogen sulfide (H₂S) production in enteric bacteria. The formulation supports identification and differentiation of organisms such as Salmonella and Shigella during microbiological analysis. The 9mL slant format provides extended media volume for laboratories that require longer incubation periods or larger inoculum s. Characteristic color changes and precipitate formation allow microbiologists to observe metabolic reactions and identify bacterial species. Packaged in sterile glass tubes, LIA slants provide a reliable and convenient solution for biochemical testing, bacterial differentiation, and routine microbiology workflows in clinical, food safety, and environmental laboratories.