Learn More
Description
- Impact-resistant 15 mL tubes contain 1.6 mm aluminum oxide particles and 1.6 mm silicon carbide particles
- With white caps
- Specific Gravity: Aluminum Oxide, 3.95; Silicon Carbide, 3.20
- Nucleic Acid Isolation and Purification
Specifications
Specifications
| Capacity | 15 mL |
| Format | Tube |
| Isolation Technology | Mechanical Disruption |
| Sample Type | Bacteria (Gram +/-), Animal Tissues, Fungi, Plants, Coral Emulsions, Mold |
| DNase | Dnase-free |
| For Use With (Application) | Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis |
| Matrix | Lysing matrix F |
| Molecular Formula | Aluminum Oxide, Al2O3; Silicon Carbide, SiC |
| Product Type | Lysing Matrix |
| Quantity | 50 x 15 mL |
| Show More |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.