Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Lysing Matrix S, 96-Tube Rack

Catalog No. MP116925001 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116925001 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116925001 Supplier MP Biomedicals™ Supplier No. 116925001
Add to Cart
Add to Cart

Lysing Matrix S contains four 1/8 in. (3.175 mm) diameter stainless steel beads. Applications include RNA and protein extraction from difficult samples where lysis by impaction with minimal mechanical shearing, is preferable.

  • Product Overview: Each impact-resistant 1.2 mL well contains four (4) 1/8 in. (3.175 mm) diameter stainless steel beads. Use to break hard/brittle samples. Specialty applications include RNA and protein extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing.
  • Applications: Metal Bead Lysing Matrix is used to break hard/brittle samples. Specialty applications include RNA and protein extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing.
TRUSTED_SUSTAINABILITY

Specifications

DNase Dnase-free
For Use With (Equipment) FastPrep-96
For Use With (Application) Sample Preparation, Sample lysis, Nucleic Acid isolation and purification, Molecular biology, Gene expression, Microbiological analysis, Gene function, Gene regulation
Matrix Lysing Matrix S
Quantity 1 Ea.
RNase Rnase-free
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.