Learn More
Bachem H-Lys(DL-2-amino-2-carboxyethyl)-OH · 2 HCl
SDP
Supplier:  Bachem 4008365.0050
Encompass_Preferred
Lysinoalanine · 2 HCl (diastereomeric mixture: LL + LD) , H-Lys(DL-2-amino-2-carboxyethyl)-OH · 2 HCl, 4418-81-9, C9H19N3O4 · 2 HCl, Mr 306.19, 50mg. LAL · 2 HCl. LAL was first observed in various proteins after alkali treatment or heating. The LL-isomer is a constituent of the lantibiotics ancovenin, cinnamicin, and duramycin. Synthetic lysinoalanine was found to have a strong chelating ability for metals. CAS-Number (net): 18810-04-3.
Catalog No. 50-259-118