Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bioworld Lysis buffer, 0.5% Tween 20, pH 8.0, 1 L

Catalog No. 50253079
Encompass_Preferred

Lysis buffer, 0.5% Tween 20, pH 8.0, 1 L

Catalog No. 50-253-079 Supplier Bioworld Supplier No. 220400692
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lysis buffer, 0.5% Tween 20, pH 8.0, 1 L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.