Learn More
MyBioSource Lysis Buffer
SDP
Catalog No. NC1860854
Encompass_Preferred
MyBioSource Lysis Buffer is a reagent used to lyse cells and extract proteins, DNA, and RNA. It contains nonionic detergents, salts, and stabilizing agents. This lysis buffer is compatible with many downstream applications including Western blot, PCR, and enzyme assays. It can lyse various sample types including mammalian cells, bacteria, yeast, insects, and tissues. The buffer is provided as a 10X concentrated solution that should be diluted before use. It is non-toxic and stable at room temperature. This is a high quality buffer from a reliable supplier.
Catalog No. NC1860854 Supplier MyBioSource Supplier No. MBS2090451100ML
MyBioSource Lysis Buffer is a reagent used to lyse cells and extract proteins, DNA, and RNA. It contains nonionic detergents, salts, and stabilizing agents. This lysis buffer is compatible with many downstream applications including Western blot, PCR, and enzyme assays. It can lyse various sample types including mammalian cells, bacteria, yeast, insects, and tissues. The buffer is provided as a 10X concentrated solution that should be diluted before use. It is non-toxic and stable at room temperature. This is a high quality buffer from a reliable supplier.