Learn More
Bioworld Lysis Buffer (Bacillus), 100 mL
Catalog No. 50253039
Encompass_Preferred
>b>Enzymatic lysis of gram-negative bacteria. Gram-negative bacteria are less susceptible than gram-positive bacteria to lysoyme and detergents. Enzymatic lysis of gram-negative bacteria requires the outer membrane to be permeabilized to expose the peptidoglycan layer to attack. The Bacillus lysis buffer kit provides the buffer and additive needed for the enzymatic lysis of gram-negative bacteria.
Catalog No. 50-253-039 Supplier Bioworld Supplier No. 220400421
>b>Enzymatic lysis of gram-negative bacteria. Gram-negative bacteria are less susceptible than gram-positive bacteria to lysoyme and detergents. Enzymatic lysis of gram-negative bacteria requires the outer membrane to be permeabilized to expose the peptidoglycan layer to attack. The Bacillus lysis buffer kit provides the buffer and additive needed for the enzymatic lysis of gram-negative bacteria.