Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ Lysis Buffer, FS

Catalog No. 501148053
Encompass_Preferred
Change view
Click to view available options
Quantity:
7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-114-8053 7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-114-8053 Supplier Applied Biosystems™ Supplier No. 4480724
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lysis Buffer, FS is designed specifically to lyse microbial organisms from food and environmental samples and prepare the microbial DNA for subsequent downstream analysis by real-time PCR.

Lysis Buffer, FS is designed specifically to lyse microbial organisms from food and environmental samples and prepare the microbial DNA for subsequent downstream analysis by real-time PCR.

This lysis buffer can be used in conjunction with Proteinase K, FS in the Pathatrix™ Auto Instrument or RapidFinder™ Direct Lysis workflows.

When used as a part of the Pathatrix™ Auto Instrument workflow, one bottle of Lysis Buffer, FS is sufficient for 700 reactions.

When used as a part of the RapidFinder™ Direct Lysis workflow, one bottle of Lysis Buffer, FS is sufficient for 100 reactions.

Related Links:

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4°C
Shipping Condition Room Temperature
Form Liquid
Product Line Pathatrix
Quantity 7 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.