Learn More
Bioworld Cell Lysis Buffer II for ChIP, liquid, ph 8.1, 250ml
Catalog No. NC3362578
Encompass_Preferred
10 mM Tris-HCL, pH to 8.1, 10 mM NaCl, 0.2% NP-40. Prepared in 3D water and filtered through 0.22 � filter. Shipped at room temperature, store at 2-8 C.
Catalog No. NC3362578 Supplier Bioworld Supplier No. 104500542
10 mM Tris-HCL, pH to 8.1, 10 mM NaCl, 0.2% NP-40. Prepared in 3D water and filtered through 0.22 � filter. Shipped at room temperature, store at 2-8 C.