Learn More
Macherey-Nagel MACHEREY-NAGEL Lysis Buffer LYS-EF, Endotoxin-Free Plasmid Isolation, Compatible With NucleoBond Xtra Kits, 1000 mL
Catalog No. NC2191118
Encompass_Preferred
The MACHEREY-NAGEL Lysis Buffer LYS-EF is designed for effective cell lysis in endotoxin-free plasmid DNA isolation, essential for high-quality DNA suitable for sensitive molecular biology applications. This buffer is particularly compatible with NucleoBond Xtra Midi and Maxi EF kits, setting it apart for its specialized formulation that meets rigorous research standards.
This item is not returnable. View return policy
Catalog No. NC2191118 Supplier Macherey-Nagel Supplier No. 740387.1000
The MACHEREY-NAGEL Lysis Buffer LYS-EF is designed for effective cell lysis in endotoxin-free plasmid DNA isolation, essential for high-quality DNA suitable for sensitive molecular biology applications. This buffer is particularly compatible with NucleoBond Xtra Midi and Maxi EF kits, setting it apart for its specialized formulation that meets rigorous research standards.