Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Lysis Matrix S, 50mL Tubes

Catalog No. MP116943010 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 x 50 mL
50 x 50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116943010 10 x 50 mL
MP116943050 50 x 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP116943010 Supplier MP Biomedicals™ Supplier No. 116943010
Add to Cart
Add to Cart

MP Biomedicals™ Lysis Matrix S Tubes are for extractions of nucleic acids, proteins, metabolites and other small molecules from tough, hard or brittle cell membranes including organisms with tough or dense exterior matrix

  • Each impact-resistant 2mL tube contains 3.175mm (0.125 in.) diameter stainless steel beads
  • Use to break hard/brittle samples
  • Specialty applications include RNA and protein extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing

Certifications

DNase and RNase free

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room temperature (15° - 30°C)
Sample Type All Samples, Bone, Fixed Samples, Food, Heart, Lung, Plants, Roots, Seeds, Skeletal Muscle, Spores, Tough Tissues
For Use With (Equipment) FastPrep-24, FastPrep 120, FastPrep-96
For Use With (Application) Sample preparation, Sample lysis, Nucleic acid isolation and purification, Molecular biology, Gene expression, Microbiological analysis; Gene function, Gene regulation
Matrix S
Product Type Lysis Matrix
Quantity 10 x 50 mL
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.