Learn More
Bioworld Lysis solution NaOH/SDS, 500 mL
Catalog No. 50253040
Encompass_Preferred
- Ultra Pure Grade - Contains 0.2 N NaOH and 1.0% SDS. Conductivity (10%, Water) @25C(umhos)- Report Titration(g/L)- 7.6-8.4
Catalog No. 50-253-040 Supplier Bioworld Supplier No. 220400431
- Ultra Pure Grade - Contains 0.2 N NaOH and 1.0% SDS. Conductivity (10%, Water) @25C(umhos)- Report Titration(g/L)- 7.6-8.4