Learn More
G-Biosciences Lysis, Stabilization and Preserving Solution
SCI_ED
Catalog No. S04231
Change view
Click to view available options
Quantity:
50 mL
Catalog No. Quantity
S04231 50 mL
1 options
Catalog No. S04231 Supplier G-Biosciences Supplier No. 7862194
Add to Cart
Add to Cart
Lysis, Stabilization and Preserving (LSP) solution is suitable to lyse biological samples and release nucleic acids and proteins.
LSP solution is suitable to lyse biological samples and release nucleic acids and proteins. It is also useful to preserve DNA, RNA and Proteins for any biological sample analysis. It preserves the genetic integrity and expression profiles of samples at ambient temperatures and completely inactivates infectious agents (viruses, bacteria, fungi, and parasites).
Specifications
Product Type Cell Lysis Buffer
Form Liquid
Quantity 50 mL