Learn More
INDICAL INC IndiMag 2 PW Lysis Strips (672)
SDP
Catalog No. NC3371151
Encompass_Preferred
The IndiMag 2 PW Lysis Strips (672) are designed specifically for the IndiMag 2 and allow the simultaneous lysis of 8 samples per strip, with volumes ranging from 100 µl to 1000 µl per sample. Each strip is capable of handling 8 samples at once and features ridges to ensure correct orientation during extraction (ridges must face left). Only lysis strips provided by INDICAL should be used to ensure compatibility with the IndiMag 2, as other plasticware may not be compatible and could cause issues with the extraction process. These strips ensure efficient and reliable sample lysis, which is crucial for optimal nucleic acid extraction.
Catalog No. NC3371151 Supplier INDICAL INC Supplier No. PW950215
The IndiMag 2 PW Lysis Strips (672) are designed specifically for the IndiMag 2 and allow the simultaneous lysis of 8 samples per strip, with volumes ranging from 100 µl to 1000 µl per sample. Each strip is capable of handling 8 samples at once and features ridges to ensure correct orientation during extraction (ridges must face left). Only lysis strips provided by INDICAL should be used to ensure compatibility with the IndiMag 2, as other plasticware may not be compatible and could cause issues with the extraction process. These strips ensure efficient and reliable sample lysis, which is crucial for optimal nucleic acid extraction.